Assay Details
| Gene Symbol: Spsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004863654.2 | XP_004863711.1 | 3 | 1097 | 116 | |
| XM_021243519.1 | XP_021099178.1 | 3 | 1017 | 116 | ||
More Information
Set Membership: |
|

Spsb1
Genomic Map