Assay Details
| Gene Symbol: Cx3cr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835228.3 | XP_004835285.2 | 2 | 1064 | 110 | |
| XM_021259689.1 | XP_021115348.1 | 2 | 1335 | 110 | ||
More Information
Set Membership: |
|

Cx3cr1
Genomic Map