Assay Details
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865872.2 | XP_004865929.1 | 1 | 182 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004865872.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
