Assay Details
| Gene Symbol: Sap30bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860694.3 | XP_004860751.2 | 11 | 973 | 87 | |
| XM_004860695.3 | XP_004860752.2 | 11 | 924 | 87 | ||
More Information
Set Membership: |
|

Sap30bp
Genomic Map