Assay Details
| Gene Symbol: LOC101700069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068026.2 | XP_012923480.1 | 1 | 244 | 85 | |
| XM_021249482.1 | XP_021105141.1 | 1 | 247 | 85 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013068026.2, XM_021249482.1 transcripts.
Set Membership: |
|

LOC101700069
Genomic Map