Assay Details
| Gene Symbol: Dnaja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838391.3 | XP_004838448.1 | 9 | 1907 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaja1| Gene Symbol: Dnaja1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838391.3 | XP_004838448.1 | 9 | 1907 | 115 | |
Set Membership: |
|