Assay Details
| Gene Symbol: Pycr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837822.3 | XP_004837879.1 | 4 - 5 | 606 | 62 | |
| XM_021250997.1 | XP_021106656.1 | 3 - 4 | 436 | 62 | ||
More Information
Set Membership: |
|

Pycr3
Genomic Map