Assay Details
| Gene Symbol: Ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838237.2 | XP_004838294.1 | 3 - 4 | 503 | 71 | |
| XM_021252789.1 | XP_021108448.1 | 3 - 4 | 506 | 71 | ||
| XM_021252790.1 | XP_021108449.1 | 3 - 4 | 313 | 71 | ||
More Information
Set Membership: |
|

Ak3
Genomic Map