Assay Details
Gene Symbol: Zfhx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004842035.2 | XP_004842092.1 | 3 - 4 | 3516 | 58 | |
XM_013071215.2 | XP_012926669.1 | 3 - 4 | 3516 | 58 | ||
XM_021256007.1 | XP_021111666.1 | 3 - 4 | 3589 | 58 | ||
XM_021256008.1 | XP_021111667.1 | 3 - 4 | 3218 | 58 | ||
XM_021256009.1 | XP_021111668.1 | 2 - 3 | 3091 | 58 | ||
XM_021256010.1 | XP_021111669.1 | 3 - 4 | 3438 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
