Assay Details
| Gene Symbol: Ets2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842369.3 | XP_004842426.1 | 1 - 2 | 357 | 68 | |
| XM_004842371.3 | XP_004842428.1 | 2 - 3 | 318 | 68 | ||
| XM_013071319.2 | XP_012926773.1 | 2 - 3 | 384 | 68 | ||
| XM_021256243.1 | XP_021111902.1 | 3 - 4 | 489 | 68 | ||
| XM_021256244.1 | XP_021111903.1 | 1 - 2 | 361 | 68 | ||
More Information
Set Membership: |
|

Ets2
Genomic Map