Assay Details
| Gene Symbol: Kcnj6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842378.2 | XP_004842435.1 | 1 - 2 | 293 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004842378.2 transcripts.
Set Membership: |
|

Kcnj6
Genomic Map