Assay Details
| Gene Symbol: Nek6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004849398.3 | XP_004849455.1 | 3 - 4 | 450 | 56 | |
| XM_004849399.3 | XP_004849456.1 | 3 - 4 | 384 | 56 | ||
| XM_021262091.1 | XP_021117750.1 | 3 - 4 | 282 | 56 | ||
| XM_021262092.1 | XP_021117751.1 | 3 - 4 | 282 | 56 | ||
More Information
Set Membership: |
|

Nek6
Genomic Map