Assay Details
| Gene Symbol: LOC101715007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853860.3 | XP_004853917.1 | 39 - 40 | 7594 | 73 | |
| XM_004853861.3 | XP_004853918.1 | 39 - 40 | 7481 | 73 | ||
| XM_004853864.3 | XP_004853921.1 | 38 - 39 | 7328 | 73 | ||
| XM_004853866.3 | XP_004853923.1 | 38 - 39 | 7295 | 73 | ||
| XM_004853867.3 | XP_004853924.1 | 46 - 47 | 7452 | 73 | ||
| XM_004853869.3 | XP_004853926.1 | 37 - 38 | 7142 | 73 | ||
| XM_004853871.3 | XP_004853928.1 | 43 - 44 | 7184 | 73 | ||
| XM_004853872.3 | XP_004853929.1 | 44 - 45 | 6989 | 73 | ||
| XM_013075467.2 | XP_012930921.1 | 46 - 47 | 7435 | 73 | ||
| XM_021265782.1 | XP_021121441.1 | 39 - 40 | 7630 | 73 | ||
| XM_021265786.1 | XP_021121445.1 | 39 - 40 | 7450 | 73 | ||
| XM_021265789.1 | XP_021121448.1 | 44 - 45 | 7140 | 73 | ||
More Information
Set Membership: |
|

LOC101715007
Genomic Map