Assay Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853968.3 | XP_004854025.1 | 1 - 2 | 189 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004853968.3 transcripts.
Set Membership: |
|

Mrps21
Genomic Map