Assay Details
| Gene Symbol: Wipf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857700.3 | XP_004857757.1 | 4 - 5 | 638 | 64 | |
| XM_004857701.3 | XP_004857758.1 | 4 - 5 | 559 | 64 | ||
| XM_004857702.3 | XP_004857759.1 | 4 - 5 | 535 | 64 | ||
| XM_021238541.1 | XP_021094200.1 | 5 - 6 | 646 | 64 | ||
More Information
Set Membership: |
|

Wipf1
Genomic Map