Assay Details
| Gene Symbol: Mlx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859321.3 | XP_004859378.1 | 6 - 7 | 618 | 62 | |
| XM_004859322.3 | XP_004859379.1 | 5 - 6 | 530 | 62 | ||
| XM_013077729.2 | XP_012933183.1 | 5 - 6 | 840 | 62 | ||
| XM_013077730.2 | XP_012933184.1 | 6 - 7 | 607 | 62 | ||
More Information
Set Membership: |
|

Mlx
Genomic Map