Assay Details
| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860186.3 | XP_004860243.2 | 10 - 11 | 1615 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man1a1| Gene Symbol: Man1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860186.3 | XP_004860243.2 | 10 - 11 | 1615 | 56 | |
Set Membership: |
|