Assay Details
| Gene Symbol: LOC101714689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860506.1 | XP_004860563.1 | 2 - 3 | 201 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101714689| Gene Symbol: LOC101714689 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860506.1 | XP_004860563.1 | 2 - 3 | 201 | 81 | |