Assay Details
| Gene Symbol: Epb42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861664.2 | XP_004861721.1 | 4 - 5 | 623 | 58 | |
| XM_021241745.1 | XP_021097404.1 | 3 - 4 | 1145 | 58 | ||
More Information
Set Membership: |
|

Epb42
Genomic Map