Assay Details
| Gene Symbol: Pbxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871331.3 | XP_004871388.1 | 4 - 5 | 542 | 59 | |
| XM_004871332.3 | XP_004871389.1 | 5 - 6 | 557 | 59 | ||
| XM_004871333.3 | XP_004871390.1 | 5 - 6 | 562 | 59 | ||
| XM_021249268.1 | XP_021104927.1 | 4 - 5 | 684 | 59 | ||
| XM_021249269.1 | XP_021104928.1 | 4 - 5 | 874 | 59 | ||
| XM_021249270.1 | XP_021104929.1 | 5 - 6 | 747 | 59 | ||
| XM_021249271.1 | XP_021104930.1 | 5 - 6 | 627 | 59 | ||
| XM_021249272.1 | XP_021104931.1 | 5 - 6 | 622 | 59 | ||
| XM_021249273.1 | XP_021104932.1 | 4 - 5 | 547 | 59 | ||
More Information
Set Membership: |
|

Pbxip1
Genomic Map