Assay Details
| Gene Symbol: Chrna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068247.2 | XP_012923701.1 | 4 - 5 | 634 | 77 | |
| XM_021250074.1 | XP_021105733.1 | 5 - 6 | 741 | 77 | ||
| XM_021250075.1 | XP_021105734.1 | 4 - 5 | 582 | 77 | ||
| XM_021250076.1 | XP_021105735.1 | 5 - 6 | 602 | 77 | ||
More Information
Set Membership: |
|

Chrna5
Genomic Map