Assay Details
| Gene Symbol: Pigz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255182.1 | XP_021110841.1 | 1 - 2 | 664 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021255182.1 transcripts.
Set Membership: |
|

Pigz
Genomic Map