Assay Details
| Gene Symbol: Pigz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255182.1 | XP_021110841.1 | 2 - 3 | 884 | 99 | |
| XM_021255185.1 | XP_021110844.1 | 2 - 3 | 422 | 99 | ||
More Information
Set Membership: |
|

Pigz
Genomic Map