Assay Details
| Gene Symbol: Tekt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263318.1 | XP_021118977.1 | 3 - 4 | 535 | 58 | |
| XM_021263319.1 | XP_021118978.1 | 3 - 4 | 535 | 58 | ||
More Information
Set Membership: |
|

Tekt2
Genomic Map