Assay Details
| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836652.3 | XP_004836709.1 | 1 - 2 | 194 | 72 | |
| XM_021241823.1 | XP_021097482.1 | 1 - 2 | 214 | 72 | ||
| XM_021241826.1 | XP_021097485.1 | 1 - 2 | 215 | 72 | ||
More Information
Set Membership: |
|

Hint1
Genomic Map