Assay Details
| Gene Symbol: Hint1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836652.3 | XP_004836709.1 | 3 - 4 | 422 | 73 | |
| XM_004836653.3 | XP_004836710.1 | 2 - 3 | 292 | 73 | ||
More Information
Set Membership: |
|

Hint1
Genomic Map