Assay Details
| Gene Symbol: Zdhhc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854860.3 | XP_004854917.1 | 10 - 11 | 1040 | 67 | |
| XM_013075839.2 | XP_012931293.1 | 11 - 12 | 1080 | 67 | ||
| XM_021236363.1 | XP_021092022.1 | 11 - 12 | 1082 | 67 | ||
| XM_021236364.1 | XP_021092023.1 | 10 - 11 | 960 | 67 | ||
| XM_021236365.1 | XP_021092024.1 | 10 - 11 | 977 | 67 | ||
More Information
Set Membership: |
|

Zdhhc20
Genomic Map