Assay Details
| Gene Symbol: Nfe2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859582.2 | XP_004859639.1 | 4 - 5 | 1517 | 59 | |
| XM_004859584.2 | XP_004859641.1 | 5 - 6 | 1226 | 59 | ||
| XM_004859585.2 | XP_004859642.1 | 4 - 5 | 1481 | 59 | ||
| XM_004859588.3 | XP_004859645.1 | 3 - 4 | 748 | 59 | ||
| XM_004859591.3 | XP_004859648.1 | 3 - 4 | 874 | 59 | ||
| XM_021240374.1 | XP_021096033.1 | 3 - 4 | 1296 | 59 | ||
More Information
Set Membership: |
|

Nfe2l1
Genomic Map