Assay Details
| Gene Symbol: LOC101698172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004860706.3 | XP_004860763.1 | 3 - 4 | 487 | 60 | |
| XM_013078181.2 | XP_012933635.1 | 3 - 4 | 487 | 60 | ||
| XM_013078182.2 | XP_012933636.1 | 3 - 4 | 487 | 60 | ||
More Information
Set Membership: |
|

LOC101698172
Genomic Map