Assay Details
| Gene Symbol: Rel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867239.3 | XP_004867296.1 | 9 - 10 | 1151 | 86 | |
| XM_021246282.1 | XP_021101941.1 | 8 - 9 | 1402 | 86 | ||
| XM_021246283.1 | XP_021101942.1 | 8 - 9 | 979 | 86 | ||
More Information
Set Membership: |
|

Rel
Genomic Map