Assay Details
| Gene Symbol: Alkal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868303.3 | XP_004868360.1 | 3 - 4 | 930 | 75 | |
| XM_021247269.1 | XP_021102928.1 | 2 - 3 | 876 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alkal2| Gene Symbol: Alkal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868303.3 | XP_004868360.1 | 3 - 4 | 930 | 75 | |
| XM_021247269.1 | XP_021102928.1 | 2 - 3 | 876 | 75 | ||