Assay Details
| Gene Symbol: Alkal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868303.3 | XP_004868360.1 | 4 - 5 | 1004 | 59 | |
| XM_021247269.1 | XP_021102928.1 | 3 - 4 | 950 | 59 | ||
More Information
Set Membership: |
|

Alkal2
Genomic Map