Assay Details
| Gene Symbol: CUNH19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872446.2 | XP_004872503.1 | 2 - 3 | 108 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004872446.2 transcripts.
Set Membership: |
|

CUNH19orf57
Genomic Map