Assay Details
| Gene Symbol: CUNH19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872446.2 | XP_004872503.1 | 4 - 5 | 357 | 103 | |
| XM_013068399.2 | XP_012923853.1 | 4 - 5 | 322 | 103 | ||
| XM_013068400.2 | XP_012923854.1 | 5 - 6 | 430 | 103 | ||
| XM_013068401.2 | XP_012923855.1 | 5 - 6 | 418 | 103 | ||
| XM_021250318.1 | XP_021105977.1 | 6 - 7 | 718 | 103 | ||
| XM_021250319.1 | XP_021105978.1 | 6 - 7 | 690 | 103 | ||
| XM_021250320.1 | XP_021105979.1 | 4 - 5 | 261 | 103 | ||
| XM_021250321.1 | XP_021105980.1 | 3 - 4 | 153 | 103 | ||
| XM_021250323.1 | XP_021105982.1 | 4 - 5 | 377 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250318.1, XM_021250319.1, XM_021250320.1, XM_021250321.1 transcripts.
Set Membership: |
|

CUNH19orf57
Genomic Map