Assay Details
| Gene Symbol: CUNH19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872446.2 | XP_004872503.1 | 6 - 7 | 1652 | 71 | |
| XM_013068399.2 | XP_012923853.1 | 6 - 7 | 1617 | 71 | ||
| XM_013068400.2 | XP_012923854.1 | 7 - 8 | 1725 | 71 | ||
| XM_013068401.2 | XP_012923855.1 | 7 - 8 | 1713 | 71 | ||
| XM_021250318.1 | XP_021105977.1 | 8 - 9 | 2013 | 71 | ||
| XM_021250319.1 | XP_021105978.1 | 8 - 9 | 1985 | 71 | ||
| XM_021250320.1 | XP_021105979.1 | 6 - 7 | 1556 | 71 | ||
| XM_021250321.1 | XP_021105980.1 | 5 - 6 | 1448 | 71 | ||
| XM_021250323.1 | XP_021105982.1 | 6 - 7 | 1672 | 71 | ||
More Information
Set Membership: |
|

CUNH19orf57
Genomic Map