Assay Details
| Gene Symbol: CUNH19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872446.2 | XP_004872503.1 | 7 - 8 | 1747 | 91 | |
| XM_013068399.2 | XP_012923853.1 | 7 - 8 | 1712 | 91 | ||
| XM_013068400.2 | XP_012923854.1 | 8 - 9 | 1820 | 91 | ||
| XM_013068401.2 | XP_012923855.1 | 8 - 9 | 1808 | 91 | ||
| XM_021250318.1 | XP_021105977.1 | 9 - 10 | 2108 | 91 | ||
| XM_021250319.1 | XP_021105978.1 | 9 - 10 | 2080 | 91 | ||
| XM_021250320.1 | XP_021105979.1 | 7 - 8 | 1651 | 91 | ||
| XM_021250321.1 | XP_021105980.1 | 6 - 7 | 1543 | 91 | ||

CUNH19orf57
Genomic Map