Assay Details
| Gene Symbol: Cd3eap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873188.3 | XP_004873245.2 | 2 - 3 | 373 | 81 | |
| XM_021250841.1 | XP_021106500.1 | 2 - 3 | 242 | 81 | ||
More Information
Set Membership: |
|

Cd3eap
Genomic Map