Assay Details
| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066562.2 | XP_012922016.2 | 7 - 8 | 960 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magt1| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013066562.2 | XP_012922016.2 | 7 - 8 | 960 | 72 | |
Set Membership: |
|