Assay Details
| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246642.1 | XP_021102301.1 | 2 - 3 | 851 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax9| Gene Symbol: Pax9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246642.1 | XP_021102301.1 | 2 - 3 | 851 | 80 | |
Set Membership: |
|