Assay Details
| Gene Symbol: Pak5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840815.3 | XP_004840872.1 | 4 - 5 | 1417 | 59 | |
| XM_021255058.1 | XP_021110717.1 | 5 - 6 | 1498 | 59 | ||
More Information
Set Membership: |
|

Pak5
Genomic Map