Assay Details
| Gene Symbol: Gatc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843663.3 | XP_004843720.1 | 2 - 3 | 314 | 85 | |
| XM_021257351.1 | XP_021113010.1 | 2 - 3 | 353 | 85 | ||
More Information
Set Membership: |
|

Gatc
Genomic Map