Assay Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848426.2 | XP_004848483.1 | 3 - 4 | 831 | 65 | |
| XM_004848427.3 | XP_004848484.1 | 3 - 4 | 812 | 65 | ||
| XM_021261334.1 | XP_021116993.1 | 3 - 4 | 798 | 65 | ||
More Information
Set Membership: |
|

Fzd3
Genomic Map