Assay Details
| Gene Symbol: CUNH1orf162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853721.2 | XP_004853778.1 | 4 - 5 | 6151 | 86 | |
| XM_021265663.1 | XP_021121322.1 | 4 - 5 | 6151 | 86 | ||
More Information
Set Membership: |
|

CUNH1orf162
Genomic Map