Assay Details
| Gene Symbol: LOC106007594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854488.3 | XP_004854545.1 | 2 - 3 | 1390 | 77 | |
| XM_021266279.1 | XP_021121938.1 | 2 - 3 | 1364 | 77 | ||
More Information
Set Membership: |
|

LOC106007594
Genomic Map