Assay Details
| Gene Symbol: LOC101726067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855792.3 | XP_004855849.1 | 20 - 21 | 2381 | 94 | |
| XM_004855793.3 | XP_004855850.1 | 20 - 21 | 2382 | 94 | ||
| XM_021237241.1 | XP_021092900.1 | 20 - 21 | 2382 | 94 | ||
More Information
Set Membership: |
|

LOC101726067
Genomic Map