Assay Details
| Gene Symbol: Chst8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858912.2 | XP_004858969.1 | 2 - 3 | 577 | 64 | |
| XM_004858913.2 | XP_004858970.1 | 4 - 5 | 520 | 64 | ||
| XM_021239722.1 | XP_021095381.1 | 2 - 3 | 782 | 64 | ||
More Information
Set Membership: |
|

Chst8
Genomic Map