Assay Details
| Gene Symbol: Slc23a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864560.3 | XP_004864617.1 | 11 - 12 | 1831 | 61 | |
| XM_004864561.3 | XP_004864618.1 | 9 - 10 | 1467 | 61 | ||
More Information
Set Membership: |
|

Slc23a3
Genomic Map