Assay Details
| Gene Symbol: Lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866074.2 | XP_004866131.1 | 9 - 10 | 1651 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmnb2| Gene Symbol: Lmnb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866074.2 | XP_004866131.1 | 9 - 10 | 1651 | 67 | |
Set Membership: |
|