Assay Details
| Gene Symbol: Cntd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013069043.2 | XP_012924497.1 | 1 - 2 | 746 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013069043.2 transcripts.
Set Membership: |
|

Cntd2
Genomic Map