Assay Details
| Gene Symbol: Vps35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237523.1 | XP_021093182.1 | 2 - 3 | 358 | 82 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps35l| Gene Symbol: Vps35l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237523.1 | XP_021093182.1 | 2 - 3 | 358 | 82 | |